Jervine
Synonym(s):
(3β, 23β)-17,23-Epoxy-3-hydroxyveratraman-11-one, 11-Ketocyclopamine
Empirical Formula (Hill Notation):
C27H39NO3
CAS Number:
Molecular Weight:
425.60
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing