Imidacloprid for system suitability
Synonym(s):
Imidacloprid
Empirical Formula (Hill Notation):
C9H10ClN5O2
CAS Number:
Molecular Weight:
255.66
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing