
IL-3 human

Synonym(s):
HCGF, MCGF (Mast cell growth factor), Multi-CSF, P-cell stimulation factor, IL-3, Interleukin-3, MGC79398
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Manufacturer
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing