![Hexylresorcinol for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/172/280/8c05f3f6-bcb9-4c27-a2bb-5201d227f274/640/8c05f3f6-bcb9-4c27-a2bb-5201d227f274.png)
Hexylresorcinol for system suitability
![Hexylresorcinol for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/172/280/8c05f3f6-bcb9-4c27-a2bb-5201d227f274/640/8c05f3f6-bcb9-4c27-a2bb-5201d227f274.png)
Synonym(s):
4-Hexylresorcinol, 4-Hexyl-1,3-dihydroxybenzene
Linear Formula:
CH3(CH2)5C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
194.27
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing