Genipin
Synonym(s):
Methyl (1S,2R,6S)-2-hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate
Empirical Formula (Hill Notation):
C11H14O5
CAS Number:
Molecular Weight:
226.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing