Skip to Content
Merck
γ-Dodecalactone natural, ≥98%, FG

γ-Dodecalactone

γ-Dodecalactone natural, ≥98%, FG
Synonym(s):
(±)-4-Dodecanolide, (±)-γ-Octyl-γ-butyrolactone, (±)-Dihydro-5-octyl-2(3H)-furanone
Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing