Skip to Content
Merck
vibrant-m

Gadobutrol for peak identification

vibrant-m
Synonym(s):
Gadobutrol monohydrate
Empirical Formula (Hill Notation):
C18H31GdN4O9 · H2O
CAS Number:
Molecular Weight:
622.73

Products

Formula Weight
Manufacturer
Application
API Family
Special Grade
Showing 1-1 of 1