Skip to Content
Merck
Fmoc-Ala-OH 95%

Fmoc-Ala-OH

Fmoc-Ala-OH 95%
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-alanine, Fmoc-L-alanine, Fmoc-Ala-OH
Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing