Flurbiprofen impurity A
Synonym(s):
Biprofen, 2-(4-Biphenylyl)propionic acid, NSC 16300
Empirical Formula (Hill Notation):
C15H14O2
CAS Number:
Molecular Weight:
226.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing