Skip to Content
Merck
vibrant-m

Felodipine Related Compound A

vibrant-m
Synonym(s):
Dehydrofelodipine, Ethyl methyl 4-(2,3-dichlorophenyl)-2,6-dimethylpyridine-3,5-dicarboxylate
Empirical Formula (Hill Notation):
C18H17Cl2NO4
CAS Number:
Molecular Weight:
382.24

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1