Skip to Content
Merck
vibrant-m

F.A.M.E. Mix RM-3

vibrant-m
Synonym(s):
AOCS Reference Mixtures, Oil Reference Standard, AOCS

Products

Product Category
Brand
Product Line
Format
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing