Skip to Content
Merck
Dynorphin A porcine ≥95% (HPLC)

Dynorphin A porcine

Dynorphin A porcine ≥95% (HPLC)
Synonym(s):
Dynorphin-(1-17)
Empirical Formula (Hill Notation):
C99H155N31O23
CAS Number:
Molecular Weight:
2147.48

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing