![Drospirenone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/229/898/09897af1-f568-48d8-a62a-b3970ca51311/640/09897af1-f568-48d8-a62a-b3970ca51311.png)
Drospirenone for peak identification
![Drospirenone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/229/898/09897af1-f568-48d8-a62a-b3970ca51311/640/09897af1-f568-48d8-a62a-b3970ca51311.png)
Synonym(s):
Drospirenone, 6β,7β:15β,16β -Dimethylene-3-oxo-17α-pregn-4-ene-21,17-carbolactone, Dihydrospirorenone
Empirical Formula (Hill Notation):
C24H30O3
CAS Number:
Molecular Weight:
366.49
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing