Skip to Content
Merck
(±)-3,4-Methylenedioxyamphetamine hydrochloride ≥98% (TLC)

(±)-3,4-Methylenedioxyamphetamine hydrochloride

(±)-3,4-Methylenedioxyamphetamine hydrochloride ≥98% (TLC)
Synonym(s):
(±)-α-Methyl-1,3-benzodioxole-5-ethanamine hydrochloride, MDA hydrochloride
Empirical Formula (Hill Notation):
C10H13NO2 · HCl
CAS Number:
Molecular Weight:
215.68

Products

Product Category
Brand
Formula Weight
Purity
Research Area
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing