![Dihydroxyfumaric acid hydrate 98%](/deepweb/assets/sigmaaldrich/product/structures/178/314/28894476-3934-407c-864c-2d85d10eb7b1/640/28894476-3934-407c-864c-2d85d10eb7b1.png)
Dihydroxyfumaric acid hydrate
![Dihydroxyfumaric acid hydrate 98%](/deepweb/assets/sigmaaldrich/product/structures/178/314/28894476-3934-407c-864c-2d85d10eb7b1/640/28894476-3934-407c-864c-2d85d10eb7b1.png)
Empirical Formula (Hill Notation):
C4H4O6 · xH2O
CAS Number:
Molecular Weight:
148.07 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing