Skip to Content
Merck
vibrant-m

Desogestrel Related Compound D

vibrant-m
Synonym(s):
13-Ethyl-11-methylenegon-4-en-17-one
Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1