Skip to Content
Merck
Cyclosporine United States Pharmacopeia (USP) Reference Standard

Cyclosporine

Cyclosporine United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Cyclosporin A, Antibiotic S 7481F1, Cyclosporine
Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Special Grade
Technique