![Curcumol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/367/296/9b47232b-dc02-4697-979d-02d431c21ab7/640/9b47232b-dc02-4697-979d-02d431c21ab7.png)
Curcumol
![Curcumol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/367/296/9b47232b-dc02-4697-979d-02d431c21ab7/640/9b47232b-dc02-4697-979d-02d431c21ab7.png)
Synonym(s):
(−)-Curcumol, (3S,3aS,5S,6R,8aS)-Octahydro-3-methyl-8-methylene-5-(1-methylethyl)-6H-3a,6-epoxyazulen-6-ol
Empirical Formula (Hill Notation):
C15H24O2
CAS Number:
Molecular Weight:
236.35
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing