
Crotyl bromide

Synonym(s):
trans-1-Bromo-2-butene
Linear Formula:
CH3CH=CHCH2Br
CAS Number:
Molecular Weight:
135.00
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing