Skip to Content
Products
Cart
0
LT
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Cefoxitin sodium salt
Empirical Formula (Hill Notation):
C
16
H
16
N
3
NaO
7
S
2
CAS Number:
33564-30-6
Molecular Weight:
449.43
EC Number:
251-574-6
Products
Product Category
antibiotics (1)
Brand
Supelco (1)
Formula Weight
Physical Form
Quality Segment
Application
Reference Material Type
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C4786
Cefoxitin sodium salt,
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More