Skip to Content
Merck
Carbamazepine Related Compound B United States Pharmacopeia (USP) Reference Standard

Carbamazepine Related Compound B

Carbamazepine Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
5H-Dibenz[b,f]azepine, Iminostilbene
Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1