![Camptothecin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/123/089/e944834d-8145-4adb-9040-27d6ac64aab8/640/e944834d-8145-4adb-9040-27d6ac64aab8.png)
Camptothecin
![Camptothecin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/123/089/e944834d-8145-4adb-9040-27d6ac64aab8/640/e944834d-8145-4adb-9040-27d6ac64aab8.png)
Synonym(s):
(S)-(+)-Camptothecin
Empirical Formula (Hill Notation):
C20H16N2O4
CAS Number:
Molecular Weight:
348.35
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing