Skip to Content
Merck
Bupropion Related Compound F Pharmaceutical Secondary Standard; Certified Reference Material

Bupropion Related Compound F

Bupropion Related Compound F Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1-(3-Chlorophenyl)-1-hydroxy-2-propanone
Empirical Formula (Hill Notation):
C9H9ClO2
CAS Number:
Molecular Weight:
184.62

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1