Skip to Content
Merck
vibrant-m

Bupropion Hydrochloride Related Compound C

vibrant-m
Synonym(s):
1-(3-Chlorophenyl)-2-hydroxy-1-propanone
Empirical Formula (Hill Notation):
C9H9ClO2
CAS Number:
Molecular Weight:
184.62

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1