Skip to Content
Merck
BDE No 209 solution ~50 μg/mL (Isooctane:Toluene 50:50), analytical standard

BDE No 209 solution

BDE No 209 solution ~50 μg/mL (Isooctane:Toluene 50:50), analytical standard
Synonym(s):
Decabromodiphenyl ether solution, Decabromodiphenyl oxide
Empirical Formula (Hill Notation):
C12Br10O
CAS Number:
Molecular Weight:
959.17

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing