
Altretamine

Synonym(s):
2,4,6-Tris(dimethylamino)-1,3,5-triazine, Altretamine, Hexamethylmelamine, N,N,N′,N′,N′′,N′′-Hexamethyl-1,3,5-triazine-2,4,6-triamine
Empirical Formula (Hill Notation):
C9H18N6
CAS Number:
Molecular Weight:
210.28
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing