![α-D-Glucose 1-phosphate disodium salt hydrate ≥97% (Enzymatic Purity, anhydrous)](/deepweb/assets/sigmaaldrich/product/structures/206/329/8be52575-4784-4916-b403-dfc357dc9801/640/8be52575-4784-4916-b403-dfc357dc9801.png)
α-D-Glucose 1-phosphate disodium salt hydrate
![α-D-Glucose 1-phosphate disodium salt hydrate ≥97% (Enzymatic Purity, anhydrous)](/deepweb/assets/sigmaaldrich/product/structures/206/329/8be52575-4784-4916-b403-dfc357dc9801/640/8be52575-4784-4916-b403-dfc357dc9801.png)
Synonym(s):
α-D-Glucopyranose 1-phosphate, Cori ester
Linear Formula:
C6H11O9PNa2 · xH2O
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing