
Acetazolamide Related Compound D

Synonym(s):
5-Amino-1,3,4-thiadiazole-2-sulfonamide, 2-Amino-1,3,4-thiadazole-5-sulfonamide, NSC 22979
Empirical Formula (Hill Notation):
C2H4N4O2S2
CAS Number:
Molecular Weight:
180.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2