Skip to Content
Merck
Abacavir Related Compound D certified reference material, pharmaceutical secondary standard

Abacavir Related Compound D

Abacavir Related Compound D certified reference material, pharmaceutical secondary standard
Synonym(s):
N6-Cyclopropyl-9-{(1R,4S)-4-[(2,5-diamino-6-chloropyrimidin-4-yloxy)methyl]cyclopent-2-enyl}-9H-purine-2,6-diamine
Empirical Formula (Hill Notation):
C18H21ClN10O
CAS Number:
Molecular Weight:
428.88

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade