Skip to Content
Merck
4-Bromo-3,5-dimethylphenyl N-methylcarbamate 98%

4-Bromo-3,5-dimethylphenyl N-methylcarbamate

4-Bromo-3,5-dimethylphenyl N-methylcarbamate 98%
Linear Formula:
CH3NHCO2C6H2(CH3)2Br
CAS Number:
Molecular Weight:
258.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing