![4-Amino-6-chlorobenzene-1-3-disulfonamide British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/182/101/8392febd-4c63-4a7b-be8f-7914cc9d848e/640/8392febd-4c63-4a7b-be8f-7914cc9d848e.png)
4-Amino-6-chlorobenzene-1-3-disulfonamide
![4-Amino-6-chlorobenzene-1-3-disulfonamide British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/182/101/8392febd-4c63-4a7b-be8f-7914cc9d848e/640/8392febd-4c63-4a7b-be8f-7914cc9d848e.png)
Synonym(s):
3-Chloroaniline-4,6-disulfonamide, 4-Amino-6-chlorobenzene-1,3-disulfonamide
Linear Formula:
H2NC6H2(Cl)(SO2NH2)2
CAS Number:
Molecular Weight:
285.73
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing