
3-Dechloro Aripiprazole

Synonym(s):
7-[4-[4-(2-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, 3-Deschloroaripiprazole
Empirical Formula (Hill Notation):
C23H28ClN3O2
CAS Number:
Molecular Weight:
413.94
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing