Skip to Content
Merck
2-Methylbutyric acid analytical standard

2-Methylbutyric acid

2-Methylbutyric acid analytical standard
Synonym(s):
NSC 7304
Linear Formula:
C2H5CH(CH3)CO2H
CAS Number:
Molecular Weight:
102.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing