
2,2-difluoroethylamine hydrochloride

Empirical Formula (Hill Notation):
C2H6ClF2N
CAS Number:
Molecular Weight:
117.53
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1