
1-Stearoyl-2-arachidonoyl-sn-glycerol

Synonym(s):
1-Stearoyl-2-arachidonoyl-sn-glycerol
Empirical Formula (Hill Notation):
C41H72O5
CAS Number:
Molecular Weight:
645.01
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing