Skip to Content
Merck
1-Chloro-3,5-dimethyladamantane 95%

1-Chloro-3,5-dimethyladamantane

1-Chloro-3,5-dimethyladamantane 95%
Empirical Formula (Hill Notation):
C12H19Cl
CAS Number:
Molecular Weight:
198.73

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing