Product Name
2-Bromohexanoyl bromide 95%
Product Number
306924
Product Brand
aldrich
CAS Number
54971-26-5
Molecular Weight
257.95
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Yellow |
APPEARANCE (FORM) | Liquid |
TITRATION | 96.5 - 103.5 % |
TITRATION (METHOD) | BROMINE COMBUSTION |
PURITY (GC AREA %) | ≥ 96.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |