G006132
HPLC Analysis of Ovalbumin and BSA Charge Variants on Proteomix® SAX-NP3
application for HPLC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
technique(s)
HPLC: suitable
test parameters
column: Proteomix SAX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777219)
mobile phase: 20 mM Tris buffer, pH 8.0
gradient: 0 - 0.3 M NaCl in 15 min
flow rate: 0.5 mL/min
pressure: 1600 psi (110 bar)
detector: UV, 214 nm
suitability
application for HPLC
application(s)
life science and biopharma
pharmaceutical (small molecule)
Legal Information
Proteomix is a registered trademark of Sepax Technologies
analytical column
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service