G005610
HPLC Analysis of Withania Extract on Ascentis® Express Phenyl-Hexyl
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Ascentis Express Phenyl-Hexyl, 10 cm x 2.1 mm I.D., 2.7 μm particles (53336-U)
mobile phase: [A] water; [B] acetonitrile
gradient: 20 to 100% B in 10 min; held at 100% B for 0.5 min
flow rate: 0.3 mL/min
pressure: 160 bar (2320 psi)
column temp.: 35 °C
detector: UV, 227 nm
injection: 5 μL
sample: 20 mg/mL in water:acetonitrile (50:50); sonicate 15 minutes; filter 0.45 μm;
dilute to water:acetonitrile (80:20)
suitability
application for HPLC
application(s)
vitamins, nutraceuticals, and natural products
Analysis Note
Legal Information
Analyte
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service