Skip to Content
Merck
All Photos(3)

Documents

ZLXM0D020

Milli-Q

Elix® Electrodeionization (EDI) Module

Water purification by electrodeionization (EDI) at 20 L/hr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

material

resin (Mixed bed ion-exchange resin)

feature

removes remaining ions by electrodeionization

packaging

pkg of 1 unit

manufacturer/tradename

Elix®

parameter

20 L/hr flow rate

compatibility

for use with Elix®

Looking for similar products? Visit Product Comparison Guide

General description

The Elix® electrodeionization (EDI) module is designed to continuously deionize water following reverse osmosis (RO). The Elix® module aids in delivering superior quality pure water consistently and reliably, without any maintenance at a flow rate of 20 L/hr.

Application

The Type 2 water produced post RO-EDI treatment meets the needs of many general laboratory applications (e.g. sample preparation, buffer and reagent preparation, glassware rinsing), and as feed for equipment and instruments (e.g. autoclaves, dishwashers, weathering and stability test chambers, clinical analyzers and slide stainers, hydrogen generators and ultrapure water systems).

Features and Benefits

  • Genuine replacement part for the Elix® EDI Module - ZLXM0D020.
  • Contains mixed bed ion-exchange resin.
  • Efficiently removes remaining ions from RO-purified water to produce constant-quality pure water, regardless of feed water quality (conductivity, CO2 levels) or RO cartridge performance.
  • Eliminates the need for hazardous chemical regeneration procedures, replacement of costly resins, changing DI cartridges, or adding softeners.
  • Reduces maintenance time, ensuring low and predictable running costs.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service