Skip to Content
Merck
All Photos(2)

Key Documents

ABS1508

Sigma-Aldrich

Anti-SREBP-1

from rabbit, purified by affinity chromatography

Synonym(s):

Sterol regulatory element-binding protein 1, Class D basic helix-loop-helix protein 1, bHLHd1, Sterol regulatory element-binding transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SREBF1(6720)

General description

Sterol regulatory element-binding protein 1 (UniProt: P36956; also known as SREBP-1, Class D basic helix-loop-helix protein 1, bHLHd1, Sterol regulatory element-binding transcription factor 1) is encoded by the SREBF1 (also known as BHLHD1, SREBP1) gene (Gene ID: 6720) in human. SREBP-1 is a multi-pass membrane protein that acts as at transcriptional activator required for lipid homeostasis. Six isoforms of SREBP-1 have been described that are produced by alternative splicing. SREBP-1 is expressed in a wide variety of tissues and is most abundant in liver and adrenal gland. In fetal lung and liver it is expressed in high levels. SREBP-1A predominates in hepatoma cell lines while isoform SREBP-1C is predominantly found in liver, adrenal gland, and ovary. Both isoforms IA and IC is found in kidney, brain, white fat, and muscle. SREBP-1 regulates transcription of the LDL receptor gene as well as the fatty acid and to a lesser degree the cholesterol synthesis pathway. It binds to the sterol regulatory element 1 (SRE-1) (5′-ATCACCCCAC-3′) and has dual sequence specificity binding to both an E-box motif (5′-ATCACGTGA-3′) and to SRE-1 (5′-ATCACCCCAC-3′). SREBP-1 has two cytoplasmic domains (aa 1-487 and 567-1147; two helical domains (aa 488-508 and 548-568), and a luminal domain (aa 509-547). In the absence of sterols, it moves from the endoplasmic reticulum to the Golgi. At low cholesterol the SCAP/SREBP complex is recruited into COPII vesicles for export from the ER. In the Golgi complex SREBPs are cleaved sequentially by site-1 and site-2 protease. The first cleavage by site-1 protease occurs within the luminal loop, the second cleavage by site-2 protease occurs within the first transmembrane domain and releases the transcription factor from the Golgi membrane.

Specificity

This rabbit polyclonal antibody detects Sterol regulatory element-binding protein 1 in human.

Immunogen

KLH-conjugated linenar peptide corresponding to 14 amino acids from the N-terminal half of human Sterol regulatory element-binding protein 1. The Immunogen sequence is conserved in SREBP-1A, B, and C and in isoform 4.

Application

Anti-SREBP-1, Cat. No. ABS1508, is a rabbit polyclonal antibdody that detects Sterol regulatory element-binding protein 1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected SREBP-1 in human adrenal gland and human kidney tissues.
Research Category
Signaling

Quality

Evaluated by Western Blotting in DU145 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected SREBP-1 in 10 µg of DU145 cell lysate.

Target description

~120 kDa observed; 121.67 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Liu et al.
Bioengineered, 12(1), 7119-7130 (2021-09-22)
Our previous studies have elucidated a possible connection between long intergenic non-protein coding RNA 2570 (LINC02570) and nasopharyngeal carcinoma (NPC). However, the precise mechanism by which LINC02570 promotes NPC remains unknown. We used quantitative polymerase chain reaction (qPCR) to detect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service