Skip to Content
Merck
All Photos(1)

Key Documents

ENAH98DB7844

Enamine

5-methyl-1,4-dinitro-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
172.1
MDL number:

mp

113-114

Iupac Name

5-methyl-1,4-dinitro-1H-imidazole

Inchi Code

1S/C4H4N4O4/c1-3-4(7(9)10)5-2-6(3)8(11)12/h2H,1H3

InChI key

IHZJEUKQRREQAB-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-7885583

Melting Point

113-114

MSDS

https://www.enaminestore.com/catalog/EN300-7885583


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service