AMBH2D6EEE86
Ambeed, Inc.
Ethyl imidazo[1,2-a]pyrimidine-2-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
ethyl imidazo[1,2-a]pyrimidine-2-carboxylate
Inchi Code
1S/C9H9N3O2/c1-2-14-8(13)7-6-12-5-3-4-10-9(12)11-7/h3-6H,2H2,1H3
InChI key
SORASSFGEFQWIH-UHFFFAOYSA-N
storage temp.
Sealed in dry,2-8C
Purity
97%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/64951-06-0.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/64951-06-0.html
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service