Skip to Content
Merck
All Photos(1)

Key Documents

393940

Sigma-Aldrich

Cantharidin

98%

Synonym(s):

3a,7a-Dimethylhexahydro-4,7-epoxyisobenzofuran-1,3-dione, 7a-Dimethylhexahydro-3a,4,7-epoxyisobenzofuran, Cantharidine, Hexahydro-3a,7a-dimethyl-4,7-epoxyisobenzofuran-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12162002

Assay

98%

mp

215-217 °C (lit.)

SMILES string

C[C@]12C3CCC(O3)[C@@]1(C)C(=O)OC2=O

InChI

1S/C10H12O4/c1-9-5-3-4-6(13-5)10(9,2)8(12)14-7(9)11/h5-6H,3-4H2,1-2H3/t5-,6+,9+,10-

InChI key

DHZBEENLJMYSHQ-XCVPVQRUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of protein phosphatase 2A.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dae Hee Lee et al.
Biomaterials, 171, 34-45 (2018-04-22)
Limitation of current anti-Vascular Endothelial Growth Factor (VEGF) cancer therapy is transitory responses, inevitable relapses and its insufficient tumor-targeting. Thus, multifaceted approaches, including the development of bispecific antibodies and combination strategies targeting different pathways have been proposed as an alternative.
Chin-Chuan Su et al.
The American journal of Chinese medicine, 43(3), 581-600 (2015-05-15)
Bladder cancer is a common malignancy worldwide. However, there is still no effective therapy for bladder cancer. In this study, we investigated the cytotoxic effects of cantharidin [a natural toxin produced (pure compound) from Chinese blister beetles (Mylabrisphalerata or Mylabriscichorii)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service