Skip to Content
Merck
All Photos(1)

Documents

371637

Sigma-Aldrich

3-(Carboxymethyl)benzothiazolium bromide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BrNO2S
CAS Number:
Molecular Weight:
274.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

solid

mp

250 °C (dec.) (lit.)

SMILES string

[Br-].OC(=O)C[n+]1csc2ccccc12

InChI

1S/C9H7NO2S.BrH/c11-9(12)5-10-6-13-8-4-2-1-3-7(8)10;/h1-4,6H,5H2;1H

InChI key

UEINBJHBQWSAHA-UHFFFAOYSA-N

General description

3-(Carboxymethyl)benzothiazolium bromide, a halogenated building block, is a thiazole derivative.3-(Carboxymethyl)benzothiazolium bromide (3-(carboxymethyl)-2-methylbenzothiazolium bromide) is a benzothiazolium salt. Synthesis of 3-(carboxymethyl)benzothiazolium bromide has been reported. It can be prepared by reacting 2-methylbenzothiazole with bromoacetic acid.

Application

3-(Carboxymethyl)benzothiazolium bromide may be used in the synthesis of novel semisquarylium dye, via reaction with 3,4-dibutoxy-3-cyclobutene-1,2-dione.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aggregation behavior of water soluble bis (benzothiazolylidene) squaraine derivatives in aqueous media.
Das S, et al.
The Journal of Physical Chemistry, 100(43), 17310-17315 (1996)
A benzothiazole-based semisquarylium dye suitable for highly selective Hg2+ sensing in aqueous media.
Bae J-S, et al.
Dyes and Pigments, 83(3), 324-327 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service