ADVH14DBDA2F
AChemBlock
N1,N1-dimethylcyclobutane-1,3-diamine dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N~1~,N~1~-dimethyl-1,3-cyclobutanediamine dihydrochloride
Inchi Code
1S/C6H14N2.2ClH/c1-8(2)6-3-5(7)4-6;;/h5-6H,3-4,7H2,1-2H3;2*1H
InChI key
PTXNWDIHHGMSQQ-UHFFFAOYSA-N
storage temp.
Store at Room Tem.
Purity
97%
Country of Origin
CN
Physical Form
white solid
Product Link
https://www.achemblock.com/p49255-n1-n1-dimethylcyclobutane-1-3-diamine-dihydrochloride.html
Synonym
n1,n1-dimethylcyclobutane-1,3-diamine dihydrochloride
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.achemblock.com/p49255-n1-n1-dimethylcyclobutane-1-3-diamine-dihydrochloride.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service