HPLC Analysis of Warfarin Enantiomers on Astec® Cellulose DMP
![HPLC Analysis of Warfarin Enantiomers on Astec® Cellulose DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/396/017/09047519-803a-4327-8ca2-6b53d4975bae/800/09047519-803a-4327-8ca2-6b53d4975bae.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] acetic acid; [C] TEA; (100:0.2:0.1, A:B:C)
column temp.
25 °C
flow rate
0.5 mL/min
detector
UV, 278 nm
injection
2 μL
sample
2 mg/mL in mobile phase
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null