Merck
Reflections on clinical applications of yoga in voice therapy with MTD.
Logopedics, phoniatrics, vocology (2012-11-10)
Carmelle Moore
ABSTRACT
This paper explores the application of modified yoga techniques, as an adjunct to voice therapy, by a speech pathologist who is also a yoga teacher. Yoga practices, with effects that may be short-term, are not considered a substitute for comprehensive and integrated somatic retraining systems (such as the Alexander Technique or Feldenkrais ATM). However, when yoga is conducted emphasizing kinaesthetic and proprioceptive awareness, the client may achieve an 'awareness state' that facilitates the learning of vocal remediation techniques (for example, by more easily 'tuning in' to the subtle sensations of supralaryngeal deconstriction). Core yoga elements and clinical applications are identified. The potential benefits and considerations when using yoga as an adjunct to the treatment of muscle tension dysphonia (MTD) are explored.
Product Description
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.80 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 105 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 3.00 μm