Merck
  • Characterization of a d-Lactate Dehydrogenase from Lactobacillus fermentum JN248 with High Phenylpyruvate Reductive Activity.
Characterization of a d-Lactate Dehydrogenase from Lactobacillus fermentum JN248 with High Phenylpyruvate Reductive Activity.
Journal of food science (2017-09-08)
Lixia Chen, Yajun Bai, Tai-Ping Fan, Xiaohui Zheng, Yujie Cai
ABSTRACT
Phenyllactic acid (PLA) is a novel antimicrobial compound. A novel NADH-dependent d-lactate dehydrogenase (d-LDH), named as LF-d-LDH0653, with high phenylpyruvate (PPA) reducing activity was isolated from Lactobacillus fermentum JN248. Its optimum pH and temperature were 8.0 and 50 °C, respectively. The Michaelis-Menten constant (K A novel d-LDH with phenylpyruvate reducing activity has been isolated and identified. It could be used as a reference for improving the production of optically pure d-PLA. d-PLA has a potential for application as antimicrobial an agent in dairy industry and baking industry, pharmaceutical agent in medicine and cosmetics.