Merck
  • Increased production of soluble vascular endothelial growth factors receptor-1 in CHO-cell line by using new combination of chitosan-protein lipid nanoparticles.
Increased production of soluble vascular endothelial growth factors receptor-1 in CHO-cell line by using new combination of chitosan-protein lipid nanoparticles.
International journal of clinical and experimental medicine (2015-03-19)
Poopak Farnia, Jalaledin Ghanavi, Afshin Bahrami, Mojgan Bandehpour, Bahram Kazemi, Ali Akbar Velayati
ABSTRACT
The soluble vascular endothelial growth factor receptor-1 (VEGFR1) or sFLT-1 has important role in antiangiogenesis. In this study, the increase expression and production of sFLT-1 fragment by newly designed ChPL-NPs nanoparticles (chitosan-protein lipid) using Chinese hamster ovary cell line (CHO) was evaluated. The assessment and purification of sFLT-1 were carried out by western blotting and fast protein liquid chromatography (FPLC). Thereafter, the angiostatic effect of gene transfer of sFLT-1 in Human umbilical vein endothelial cell line (HUVEC) was evaluated. Our results showed a significance rate of transfection with ChPL-NPs (80-85%) in comparison to standard lipofectamine(2000) (65-70%) (P < 0.05). The anti-angiogenic action of sFLT-1 was observed by in-vitro culture of recombinant protein (sFLT-1; 50 ng/ml) with HUVEC cell lines (5 × 10(6)). The ChPL-NPs nanoparticles can consider a potential carrier system for large scale production of sFLT-1, which ultimately may be use as therapeutic agent in targeting solid tumor tissues.
Sigma-Aldrich
Fibroblast Growth Factor-10 human, >97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available. Contact Technical Service